Density/Slope Class Set

Supplier: Shiv Dial Sud & Sons
PH10055A
470148-774CS 2841.8 USD
470148-774
Density/Slope Class Set
Educational Materials Physics Educational Materials Mechanics Learning Activities
Educators can conduct a comprehensive array of density experiments with the density/slope class set.

  • Set of 12 materials includes: Maple wood, polypropylene, nylon, acrylic, polyurethane, phenolic, PVC, copper, brass, teflon, polyethylene, and aluminum
  • For demonstrations, group work, or individual study
  • Instruction manual with suggested activities included
  • Clear plastic containers with caps provide easy storage

Adaptable to demonstrations, student group work, or individual study, this set of 12 specimens guides students to conclude that the relationship between volume and mass is density. Each specimen is a different color and comes in four separate sizes. Students measure and weigh the samples, then create a graph of their data, which leads to the identification of the materials present and reveals the relationships between mass and volume.

The large set is provided in clear plastic containers with caps for compact storage, damage protection, and use in experiments. Two extra containers are provided.

Ordering information: This kit includes an instruction manual with suggested activities.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR