Energy Transformation Kit
Supplier: VWR International
P20360A
470148-756CS 540.3 USD
470148-756
Energy Transformation Kit
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Appropriate for grades 6 to 12 Brings the First Law of Thermodynamics to life! This pair of precision-machined, chrome-plated steel balls is actually a hands-on demonstration in energy transformation. By striking the balls together, you will create enough heat at the point of contact to burn a hole in a sheet of paper.
There will be no flame but a charred hole will appear instantly along with the smell of burning paper. Each ball weighs 454 g. Includes instructions with suggestions for tying the activity into national science standards.