Mini Density Cube Set
Supplier: OSAW INDUSTRIAL PRODUCTS SE
SW520A
470148-735CS 136.55 USD
470148-735
Mini Density Cube Set
Educational Materials Physics Educational Materials Mechanics Learning Activities
Students learn about density, mass, volume, buoyancy, specific gravity, and flotation while experimenting with real-world materials.
  • Aluminum, Copper, Steel, Brass, and Zinc Construction
  • For a Variety of Experiments
  • Ideal for Learning about Concepts of Density
  • Five Cubes Included
  • Applicable Subjects Include Mass, Volume, Buoyancy, and More
The cubes are made of aluminum, copper, steel, brass, and zinc.
The five 10 mm cubes can be used in a variety of experiments.