Acrylic Prism Set
Supplier: Shiv Dial Sud & Sons
P30277
470148-722CS 1106.8 USD
470148-722
Acrylic Prism Set
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
A must for your optics lab!
  • Set of three
Set includes equilateral prisms of crystal clear acrylic in sizes 25 mm, 50 mm, and 100 mm.