Peat Pots
Supplier: PARK SEED WHOLESALE
00624-CS-SP
470147-452CS 1122.8 USD
470147-452
Peat Pots
Trays/Dishes/Pots Pots
Composed of peat moss and wood fiber, these pots allow the seedling to be planted without removing the pot. They allow easy penetration by growing roots and decompose as the plant develops.
Packaging: Package of 24.