Fever Tester
Supplier: Liquid Crystal Resources
BFT45
470146-260CS 388.8 USD
470146-260
Fever Tester
Thermometers Analog Thermometers
Accurate Readings in Fifteen Seconds
Simply press this liquid crystal thermometer against the forehead and in 15 seconds the correct temperature is highlighted in green. No sterilization is required. Range: 95–104°F. Length: 3 1/2".