GREEN Low Cost Estuary and Marine Water Monitoring Kit
Supplier: LaMotte
5911
470146-202EA 76.75 USD
470146-202
GREEN Low Cost Estuary and Marine Water Monitoring Kit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Economical Water Testing Solution
  • No hazardous compounds
  • Can be used in the field or the lab
  • Single factor tests
Designed to provide simple, economical, and non-hazardous methods for testing saline and brackish water, this kit provides 10 tests each for pH, DO, BOD, temperature, turbidity, phosphate, chloride, and two for total coliform. All contents and chemicals are non-hazardous and field-friendly.