10-Gallon Glass Aquarium Accessories
Supplier: Pet World Inc
80510
470005-132EA 25.1 USD
470005-132 470004-732 470145-890
10-Gallon Glass Aquarium Accessories
Specimens Live Specimens Aquaria - Living Material
Hinged Glass Aquarium Top
Add the hinged top to reduce evaporation and prevent fish from jumping out of the tank. Durable polypropylene hinges allow the top to open easily for quick access to the tank; the plastic strip at the back can be cut to add accessories.
Screen Aquarium Top
It features an inset metal screen in the center to easily convert your aquarium to a terrarium. You can even add large plastic tubes at the access holes to turn it into a gerbil run. It also accommodates a light strip.
Full light Hood
This tank lid has a raised light strip built in, with electrical components sealed for safety. The inner shell will not discolor or corrode from freshwater or saltwater. Durable polypropylene hinges allow the top to open easily for quick access to the tank; the plastic strip at the back can be cut to add accessories. A fluorescent bulb and instructions are included. 120 VAC.