Boreal Science Microscope Maintenance Kit
Supplier: ALGER MACHINE CO. INC.
2291200 717720 HEK7M
470031-798EA 50.1 USD
470031-798 470145-884 470134-762
Boreal Science Microscope Maintenance Kit
Tool Sets
Keep your microscope in ideal working order with these supplies for maintenance and upkeep.
Designed specifically for our full line of microscopes, this kit helps you perform the routine maintenance on your microscopes that keeps it in perfect working order.
Delivery information: This toolbox includes one 8 oz. bottle of Boreal lens cleaning solution, one No.2 Phillips head screwdriver, one 1 oz. pipet-aid, one plastic dropper, one camel’s hair brush, one 'C' wrench for microscope tension collars, one package of lens paper, one box of small Kimwipes, one toothbrush, One 1.75 oz. tube of grease, one 3×/6× magnifier, one pair of snipe-nose pliers, one hex-wrench set (7 sizes), one precision screwdriver, one pair of straight sharp-point forceps, and one slotted screwdriver (5 cm blade). The Boreal Microscope Manual with detailed instructions and a troubleshooting guide are also included.
Packaging: Items packed in heavy-duty vinyl tool roll.