Glass coverslips
Supplier: C & A Scientific
94-22222
470145-876PK 7.8 USD
470145-876
Glass coverslips
Cover Glasses
Finest Optical Quality
  • Quality slide making supplies
  • Traditional coverslip type
These cover glasses are required when using 100X oil immersion objective lenses.