VWR® Standard Sieve Set
Supplier: Avantor
470145-666KT 647.39 USD
470145-666
VWR® Standard Sieve Set
Sieves Test Sieves
The most durable sieve set on the market.
  • Made of rust-free polished and lacuqered brass
  • 8" diameter conforms to ASTM E-11 standards
  • 2" deep
  • Openings are indicated in milimeters and inches
These sieves stack together to accommodate any type of sample.
Ordering information: Each kit contains a set of 7 brass sieves as well as bottom and cover. Sieve sizes are: 5, 10, 18, 35, 60, 120, and 230.