Plastic Droppers
Supplier: Skywill Far East Industries
2921400
470145-614BX 529.35 USD
470145-614
Plastic Droppers
Pipettes Pasteur/Transfer Pipettes
Perfect for student use.
  • Each pack contains six medicine droppers with rubber bulbs
The total length of the dropper is 99 mm and capacity of each press is approximately 0.5 ml maximum.
Caution: WARNING: CHOKING HAZARD- Small Parts. Not for Children Under 3 Years.