Boreal Science Advanced Stereo Microscopes
Supplier: VWR
5789650
470145-568EA 467.85 USD
470145-568
Boreal Science Advanced Stereo Microscopes
Microscopes
This sturdy microscope is made of solid metal and coated glass optics.
  • Three-way illumination
  • Interchangeable stage plates: Translucent and black/white
  • Inclined 360° rotating head for viewing comfort
Features include an inclined head for viewing comfort, a full-size body at 30 cm, and an easy-adjust collar for precise control of focus tension. With rack and pinion focusing with slip clutch, die-cast metal construction, and a reagent-resistant finish, this stereomicroscope is durable.
It has turret-mounted 2× and 4× parfocal objectives for quick magnification changes, an inclined head for viewing comfort, and a dual illumination system for clear specimen lighting. The paired 10× wide-field eyepieces have removable, soft eyecups to help shut out peripheral light and are mounted in eyetubes with interpupillary and diopter adjustments.
Ordering information: This microscope includes a vinyl dust cover.