Bulk Spill Supplies
Supplier: CLIFFORD W. ESTES CO., INC.
06607
470043-004PK 66.25 USD
470043-004 470145-380
Bulk Spill Supplies
Spill Control Materials
Use inert, noncombustible vermiculite to contain solvent spills, sodium bicarbonate to neutralize acidic spills and waste, or sand to smother small fires.