Bel-Art Reagent/Acid Pump Dispenser, SP Scienceware
Supplier: Bel-Art
F11656-0000
470145-338CS 1139.1 USD
470145-338
Bel-Art Reagent/Acid Pump Dispenser, SP Scienceware
Pumps Vacuum Pumps
Aids in the transfer of acids and reagents and virtually eliminates accidental bottle breakage and spills.
  • Squeeze the small plastic bottle to begin flow; press the pressure release button to immediately stop dispensing
  • Fits many 5 pint (80 fl. oz.) acid bottles, ½ and 1 gallon jugs, 2.5 and 4 liter bottles, and similar bottles with GPS No. 38-430 or 38-400 threaded necks (not included)
  • Two adapters provided for short necks