Polypropylene Scoops
Supplier: Bel-Art
F36756-0000_SE
470145-336CS 207.05 USD
470145-336
Polypropylene Scoops
Scoops
Molded of strong, smooth polypropylene.
Steam autoclavable at 120°C. Scoop size: 8 1/2" x 5" x 3". Handle Length: 4".
Delivery information: Package of six.