Magnetic Stirring Bar Assortment, Teflon® Coated
Supplier: Bel-Art
37160-6001_SE
470145-284CS 745.55 USD
470145-284
Magnetic Stirring Bar Assortment, Teflon® Coated
Stirrers Magnetic Stirring Bars
Set of Six Spinbars.
  • Octagonal shape
  • Bars have pivot rings
Set includes the following sizes (Dia x H): (2) 8 x 25 mm, (2) 8 x 38 mm, and (2) 8 x 51 mm.