Bel-Bulb Pipettor
Supplier: Bel-Art
F37887-0000_SE
470145-138EA 36.45 USD
470145-138
Bel-Bulb Pipettor
Pipettes Pipette Bulbs
A time saver that eliminates mouth pipetting!
The Bel-bulb Pipettor is simply held against the pipette end and fluid is drawn up.
When the pipette is full, the bulb is removed and the pipette end is covered with the finger to hold the fluid or the fluid can be released as necessary.
Designed to fit pipettes from 6 to 8 mm in Ø, the flexible vinyl bulb is 51 mm in Ø by 70 mm long with a soft tapered chuck.