Scoop, 250 ml, 6.4 cm Handle
Supplier: Bel-Art
F36753-0000_SE
470145-134PK 41.15 USD
470145-134
Scoop, 250 ml, 6.4 cm Handle
Scoops
Rigid and strong scoop made from white polypropylene.
  • Flat bottom keeps it upright when filled or empty
  • Steam autoclavable at 121ºC (250ºF)
Designed for retrieving samples from a larger source.
Scoop, 250mL, 6.4cm Handle