KleenGuard™ Pre-Moistened Lens Cleaning Towelettes, Kimberly-Clark Professional
Supplier: Ansell Healthcare
14551
470145-122CS 210.5 USD
470145-122
KleenGuard™ Pre-Moistened Lens Cleaning Towelettes, Kimberly-Clark Professional
Wipes Disposable Wipes and Towels Lens Papers
KleenGuard™ Personal Protective Equipment (PPE) enables rather than inhibits, allowing for maximum productivity and safety.
  • Lens cleaning wipers pre-saturated with an anti-fog, anti-static formula (does not contain silicone)
  • Packaged in a wall-mountable corrugate box designed for easy dispensing
  • Each towelette is individually packaged for cleanliness and convenience
By keeping safety glasses clean, they can be extended in their use and reduce costs. Each individually wrapped towelette is pre-moistened for convenience.
KleenGuard™ pre-moistened lens cleaning towelettes offer a convenient way to keep vital safety glasses and protective goggles clean, even on the go.
The dispenser box comes in a wall-mountable corrugate design, so it can be hung wherever is most convenient.
Note: IPA-based lens cleaning towelettes can degrade eyewear anti-fog coating over time
Packaging: 10 boxes of 100 towelettes, 1000 towelettes/case.