Colorflame Candles
Supplier: JOELSON INDUSTRIES
BBB7632-52
470145-064CS 247.7 USD
470145-064
Colorflame Candles
Educational Materials Chemistry Educational Materials Properties of Matter Learning Activities in Chemistry
Excellent candles for flame tests. Add an element of fun to your chemistry demonstrations with these colorful candles.
  • Non-toxic and smokeless
  • Biodegradable
The standard birthday candles are anything but traditional. Thes candles burn with red, purple, green, blue, orange, and white flames.
Delivery information: Includes 12 candles an 12 clear plastic holders.