Polypropylene Tubing Holders
Supplier: Bel-Art
F18235-0000
470145-012KT 51 USD
470145-012
Polypropylene Tubing Holders
Clamps Tubing and Sanitary Clamps
Autoclavable polycarbonate clip that fits over the lip of a beaker, bath, or other vessel.
The 5 cm (2") long arm extends over the beaker and secures the end of a plastic or rubber tube. The maximum size tubing accepted is 9.5 mm ( 3/8") OD.
Packaging: Package of six.