Disposable Electrodes
Supplier: BIOPAC SYSTEMS INC.
EL503-10
470144-998CS 1193.2 USD
470144-998
Disposable Electrodes
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Used in ECG, EMG, and EOG experiments, these electrodes allow the attachment of three-lead electrode cables or individual electrode lead wires onto the snap connector. They come pregelled with self-adhesive hypoallergenic foam.