Estes MAXI Launch Rod
Supplier: Estes Industries
2244
470144-978EA 20.85 USD
470144-978
Estes MAXI Launch Rod
Educational Materials Physics Educational Materials Mechanics Learning Activities
Launch rod with extra strength and length for larger rockets.
  • Compatible with Porta-Pad E and Porta-Pad II launch pads
  • Two-piece design for easier storage
Going big? You might need a stronger launch rod, like this 3/16" dia. rod built for heavier rockets and higher power motors. It attaches to the launch pad, and your Estes rocket simply slides over the rod using the launch lugs attached to the rocket. When assembled, the total length is perfect to accelerate most Estes rockets to that speed for most larger rockets that are powered by C11, D and E engines. Also works great when used as an alignment tool on model rockets that require two launch lugs.