Wide Range Hydrion Double Roll Dispenser
Supplier: Micro Essential
140
470144-894CS 172.35 USD
470144-894
Wide Range Hydrion Double Roll Dispenser
Test Paper pH Indicator Papers and Test Strips
See Dramatic Color Changes
  • Two 4.6 m rolls in one dispenser
  • Tests from 1-12 on pH scale in 1.0 units
  • Color chart included