Traditional Indoor Weather Center
Supplier: SPRINGFIELD PRECISION
91001
470144-881CS 887.75 USD
470144-881
Traditional Indoor Weather Center
Weather Stations
Attractive Display, Accurate Readings.
  • Can be mounted on an indoor wall
  • Measures multiple parameters
It features separate analog displays for indoor temperature and relative humidity, as well as barometric pressure. Brasstone bezels are surrounded by a handsome cherry finished solid wood frame for wall mounting.