Low Stender-Form Preparation Dish
Supplier: Variety Glass
679
470144-866CS 996.25 USD
470144-866
Low Stender-Form Preparation Dish
Culture Dishes Petri Dishes
Dish is made of clear pressed glass. Cover is made of plain pressed glass and ground to fit. Size: 60mm Dia. x 29mm H.
Made of flint glass. Must be heated and cooled slowly. Can be heated up to 285 degrees Celsius.