Polypropylene Griffin Graduated Beakers
Supplier: Shiv Dial Sud & Sons
CH10155/13 CH10155/9 BPO50
470144-852CS 49.89 USD
470144-852 470144-854 470144-856
Polypropylene Griffin Graduated Beakers
Beakers
Economical, Unbreakable Beakers.
  • Permanent molded graduations
  • Special no-drip spouts
  • Autoclavable up to 121°C