Plastic Trays
Supplier: United Scientific Supplies
81701
470144-822CS 294.85 USD
470144-822
Plastic Trays
Trays/Dishes/Pots Laboratory Trays/Dishes
General use tray for all laboratory needs.
  • Polypropylene
  • Autoclavable
Multi-purpose polypropylene molded trays can be used for sterilizing, drying glassware, porcelain ware, and other laboratory supplies. These steam autoclavable trays have tapered walls designed for nesting.