Extreme Temperature Gloves
Supplier: Newtex Industries
2100011
470144-720CS 1349.75 USD
470144-720
Extreme Temperature Gloves
Gloves
May be used at temperatures from 70 up to 650 °C for short periods, glove's lint-free woven Zetex™ fiber provides excellent abrasion resistance.
  • Gloves feature 10 oz. wool lining
  • Double sewn for extra strength, and have a loose fitting cuff
  • 11" long to protect the wrists
Heat resistant gloves made from ZetexPlus® Vermiculite fabric for extreme heat protection to 2000 °F.