Laboratory Magnet Set
Supplier: United Scientific Supplies
MSET4 MSET4
470144-622EA 70.45 USD
470144-622 470200-924
Laboratory Magnet Set
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
A must for the Lab
  • Set of four
The magnets in this set are permanent Alnico and include two bars, two horseshoes, and keepers. Sizes: bars 3"L x 5/8"W x 3/8"H; small horseshoe 1 "W x 1"H x 3/8"D; large horseshoe 1 3/8"W x 1 3/8"H x 3/4"D.