United Scientific™ Lamp Sockets
Supplier: United Scientific Supplies
LMPPS-PK/15
470144-544CS 358.2 USD
470144-544 470188-428
United Scientific™ Lamp Sockets
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Minature sockets for small bulbs.
  • Two terminals for easy conections
  • All-metal construction
  • Fits E10 screw base mini bulbs
These miniature metal sockets have two terminals and are compatible with all of our miniature screw base incandescent lamps. Package of 15.