Square Polystyrene Disposable, Sterile Petri Dishes
Supplier: Simport Scientific
D210-16
470144-494CS 812.3 USD
470144-494
Square Polystyrene Disposable, Sterile Petri Dishes
Culture Dishes Petri Dishes
Petri dishes having a raised square grid on the bottom part. The grid consists of 36 small squares; each square is identified to facilitate special type counting or location of specimens. Supplied with top and bottom in sterile shelf packages. Size: 100 x 15mm.