Boreal Science Digital Stereomicroscope
Supplier: National Optical
DC4-456H
470144-208CS 3620 USD
470144-208
Boreal Science Digital Stereomicroscope
Microscopes
Digital stereoscope with a video camera built into the traditional microscope chassis.
  • Inclined binocular head
  • Paired 10X widefield eyepieces
  • Interpupillary adjustment in both
  • Diopter controls in one
  • Dual 1X and 3X magnification housed in 90° rotating turret
  • 10X and 30X magnification
  • Coarse focus control
  • Two removable stage plates with clips (frosted and black/white)
Microscope features a built-in 10W tungsten for transmitted illumination, 10W halogen for reflected illumination and a three-way in-base illumination control to select reflected light, transmitted light, or both. The Boreal rugged, die-cast metal body has a high-resolution camera housed in the head, and one power supply runs both the microscope and the camera. The included user-friendly Motic image editing software lets you capture images in JPG, TIFF, MIG and BMP formats, and movies in AVI format. You can also program regular interval times to capture images, perform photo editing, record a video, measure objects, label structures, compare side-by-side or superimposed images, amalgamate images, and record audio. Connect to a computer via a USB port. Comes with all connecting cables. 100–120 VAC with a 5' 6" three-wire cord.
Ordering information: System Requirements for Mac: G4, iMac, PowerPC, OSX, 128MB RAM, 300 MB hard drive space; for Windows Pentium, Windows 98/NT/2000/XP, 128MB RAM, 300 MB hard drive space.