Electric Exogeric Device
Supplier: NADA SCIENTIFIC LTD.
N99-B-2637-015
470142-930EA 41.25 USD
470142-930
Electric Exogeric Device
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Demonstrate the exothermic effect of current in a resistor.
  • Cement resistors: 5 Ohm, 20 W
  • Liquid crystal thermometer, 10 to 56 °C
Used with the GENECON®12 to conduct exothermic experiments on current by connecting a liquid crystal thermometer with a resistor. The transformation of mechanical energy is clearly demonstrated.
Applications: Physics, thermodynamics and electricity