NeuLog Respiration Monitor Belt Logger Sensor, EISCO SCIENTIFIC LLC
Supplier: Wards
NUL236
470140-290EA 250.1 USD
470140-290
NeuLog Respiration Monitor Belt Logger Sensor, EISCO SCIENTIFIC LLC
Data Loggers
Respiration impacts heart rate.
  • Max. sample rate 100 S/sec
  • Experiment duration 1 sec. to 31 days
  • Belt included
This sensor monitors abdominal or thoracical breathing as well as giving an indication of the depth of breathing.
Ordering information: For each probe you will need one or more of the following items: Battery Module, Viewer module (not needed if connecting to a computer to display data), USB module (not needed if only using as a stand alone probe or you have the WIFI module), WIFI module (Not needed if connecting to a computer through the USB module). Recommended is to have a battery module and a USB module.
Caution: For educational use only.