Density Cylinder Set

Supplier: Wards
PH0112F
470138-850KT 33.1 USD
470138-850
Density Cylinder Set
Educational Materials Physics Educational Materials Mechanics Learning Activities
This Equal Mass Density Set is for specific gravity and density determinations.

  • For Specific Gravity and Density Determinations
  • Four Cylinders of Varying Volumes Included
  • Width of All Cylinders: 16 mm dia.
  • Consist of: Cu, Fe, Al, Zn

It's used to study the specific relationship between mass and density.

The set includes four cylinders with the same mass and diameter (16 mm) but varying volumes.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR