Somso® Equal Cleavage and Gastrulation in the Lancelet
Supplier: MARCUS SOMMER
ZOS 58
470138-240EA 720.77 USD
470138-240
Somso® Equal Cleavage and Gastrulation in the Lancelet
Models Human Biology Models
Somso® premium quality models.
  • Five year warranty
Branchiostoma lanceolatum, enlarged approx. 500 times, in SOMSO-Plast®. Nine models (A - J) on stands with green bases show the various stages in cell division, the formation of the blastula and original membrane. In one piece. Weight of the series: 1.9 kg.
Ordering information: Items on this page are not stocked and may have an extended time for delivery.