Boss Head, Double Swivel
Supplier: Wards
CH0666
470137-842EA 10.65 USD
470137-842
Boss Head, Double Swivel
Clamps Bossheads
Our swiveling boss head allows you to hold two rods at any angle.
Made of die cast zinc with the ability to hold rods up to 16 mm in diameter.