Eisco® Boss Head, Aluminum
Supplier: Wards
CH0664A
470137-834EA 8.4 USD
470137-834
Eisco® Boss Head, Aluminum
Clamps Bossheads
Secure laboratory equipment and apparatus at right angles.
  • Heavy duty, aluminum die cast
  • Accepts rods up to 16 mm (Ø)
  • Holds rods at right angles
This die cast aluminum boss head holds rods securely at right angles. The metal thumbscrews are durable, easy-to-use and provide a very secure grip.