Boss Head, Square
Supplier: Wards
CH0664
470137-832EA 8.45 USD
470137-832
Boss Head, Square
Clamps Bossheads
Perfect for Physics
This square boss head accommodates both square and tubular sections up to 16mm and holds them at right angles to each other. Holds a wide variety of objects including: rules, magnets, drawing boards, and more. The alloy body and plastic thumbscrews make it perfect for any physics lab.