Solderless Breadboard
Supplier: GLOBAL SPECIALTIES SE
GS-830T
470137-790EA 7.99 USD
470137-790
Solderless Breadboard
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Low Cost Meets Quality.
  • 830 tie points
  • Size 6.5×2.1"
This solderless breadboarding system has 830 tie points, and allows students to perform lab experiments in all areas of custom electronic design applications and to design test fixtures. The color coded rails and clearly labeled grid allow for easy viewing and help students understand the conductive pathways.