Moisture Resistance Indicator
Supplier: GLOBAL SPECIALTIES SE
GSK-944
470137-772EA 14.99 USD
470137-772
Moisture Resistance Indicator
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Check the Conductivity
Use this checker to measure dissolved solids in water. The greater the resistance, the less conductive the water; as resistance levels lessen, the materials that conduct electricity are elevated.The results are displayed via LED.