Solar Fan
Supplier: GLOBAL SPECIALTIES SE
GSK-1001
470137-764EA 27.99 USD
470137-764
Solar Fan
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Makes Solar Energy Easy to Understand
Use this kit to create a circuit for converting sunlight into electrical energy. The fan speed is controlled by the intensity of the sunlight.