Tangent Galvanometer
Supplier: Wards
PH0811
470137-638EA 138.56 USD
470137-638
Tangent Galvanometer
Electrical Meters Voltmeters
This versatile galvanometer demonstrates how a current loop generates a magnetic field.
  • Five, Ten, or Fifteen Turns
  • Versatile
  • Acrylic Hoop
  • Three Binding Posts
  • Two Coils Of Copper Wire
The galvanometer includes one 12.7 cm diameter acrylic hoop wrapped with two coils of copper wire. One coil has five turns, and the other coil has ten.
Three binding posts allow connections to five, ten, or 15 turns and a compass.