Color filters
Supplier: Wards
PH0648ST8
470137-544PK
14.85
USD
470137-544
Color filters
Educational Materials
Physics Educational Materials
Optics Learning Activities
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...