NeuLog USB Module
Supplier: Wards
USB200
470137-442EA 69.85 USD
470137-442
NeuLog USB Module
Data Loggers
Easily connect your Neulog probes to your computer.
  • The USB module enables a fast connection of the sensors to the computer (PC, Mac, XO, or Linux)
This module is the first in a chain of sensors connected to a USB port of the PC. It provides both power from the PC to the sensors and communication between the PC and the sensors.
The connection to the PC is done by using a standard USB to Mini USB connecting cable (camera type), included with the USB module.
Caution: For educational use only.