NeuLog Wide-Range Temperature Logger Sensor, EISCO SCIENTIFIC LLC
Supplier: Wards
NUL234
470137-434EA 109.8 USD
470137-434
NeuLog Wide-Range Temperature Logger Sensor, EISCO SCIENTIFIC LLC
Data Loggers
Measures a wide range of temperatures.
  • Max. sample rate 100 S/sec.
  • Experimental duration 1 sec. to 31 days
Based on a thermocouple sensor, this unit enables the measurement of very high and very low temperature levels. It runs in two modes of operation either Celsius or Fahrenheit.
Ordering information: For each probe you will need one or more of the following items: Battery Module, Viewer module (not needed if connecting to a computer to display data), USB module (not needed if only using as a stand alone probe or you have the WIFI module), WIFI module (Not needed if connecting to a computer through the USB module). Recommended is to have a battery module and a USB module.
Caution: For educational use only.