NeuLog Surface Temperature Logger Sensor, EISCO SCIENTIFIC LLC
Supplier: Wards
NUL233
470137-432EA 51.59 USD
470137-432
NeuLog Surface Temperature Logger Sensor, EISCO SCIENTIFIC LLC
Data Loggers
This tough sensor is usable on any surface or even in water.
  • Two modes of operation: celsius and fahrenheit
  • Sensor works on any surface
  • Functional in water
  • Maximum sample rate of 100 S/s
It supports experiments that last between one second and 31 days.
Ordering information: The following items are needed but not included: a battery module, viewer module (not needed if connecting to a computer to display data), USB module (not needed if using as a stand-alone probe or you have the Wi-Fi module), and Wi-Fi module (not needed if connecting to aMore Product Information