NeuLog Carbon Dioxide Logger Sensor
Supplier: Wards
NUL260
470137-408EA 383.55 USD
470137-408
NeuLog Carbon Dioxide Logger Sensor
Data Loggers
This sensor monitors changes in CO2 levels.
  • Sensor Tracks CO2 Levels During Respiration
  • Measurements in ppm
  • For Experiments between One Second and 31 Days
This logger sensor measures in ppm (parts of CO2 per million of air).
This sensor supports experiments between one second and 31 days long.
Ordering information: The following items are needed but not included: a battery module, viewer module (not needed if connecting to a computer to display data), USB module (not needed if using as a stand-alone probe or you have the Wi-Fi module), and Wi-Fi module (not needed if connecting to a computer through the USB module). A battery module and a USB module are recommended.
Caution: For educational use only.